3-((6-Bromopyridin-2-yl)amino)propanoic acid 97%
Supplier: Ambeed
|
Warning
|
A115522-100MG
A115522-1G
A115522-250MG
A115522-25G
A115522-5G
77136-216EA
26.78
USD
77136-216
77136-218
77136-220
77136-222
77136-224
3-((6-Bromopyridin-2-yl)amino)propanoic acid 97%
3-((6-Bromopyridin-2-yl)amino)propanoic acid
Precaución: for Research Use Only
Formula:
C₈H₉BrN₂O₂ |
MDL Number:
MFCD27927373 CAS Number: 68638-67-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...