tert-Butyl (4-oxo-4-phenylbutyl)carbamate 97%

Supplier: Ambeed
Warning

A115386-250MG A115386-1G
77136-214EA 19.57 USD
77136-214 77570-908
tert-Butyl (4-oxo-4-phenylbutyl)carbamate 97%

Precaución: for Research Use Only

Formula: C₁₅H₂₁NO₃
MDL Number: MFCD08060130
CAS Number: 116437-41-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR