2-Chloro-3-cyano-6-picoline 98%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-3-cyano-6-methylpyridine, 2-Chloro-6-methylnicotinonitrile

A114868-100G A114868-10G A114868-1G A114868-25G A114868-5G
77136-198EA 201.58 USD
77136-198 77136-200 77136-202 77136-204 77136-206
2-Chloro-3-cyano-6-picoline 98%
2-Chloro-3-cyano-6-picoline

Precaución: for Research Use Only

Formula: C₇H₅ClN₂
MW: 152.58 g/mol
MDL Number: MFCD00082767
CAS Number: 28900-10-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR