(2R,3R)-(-)-2,3-Butanediol 98%

Supplier: Ambeed
Warning

A114648-100G A114648-10G A114648-1G A114648-25G A114648-5G
77136-186EA 593.28 USD
77136-186 77136-188 77136-190 77136-192 77136-194
(2R,3R)-(-)-2,3-Butanediol 98%
(2R,3R)-(-)-2,3-Butanediol

Precaución: for Research Use Only

Formula: C₄H₁₀O₂
MDL Number: MFCD00064267
CAS Number: 24347-58-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR