5-Iodopyrimidine-2,4-diamine 97%

Supplier: Ambeed
Warning

A114506-100MG A114506-1G
77136-184EA 14.42 USD
77136-184 77685-016
5-Iodopyrimidine-2,4-diamine 97%
5-Iodopyrimidine-2,4-diamine

Precaución: for Research Use Only

Formula: C₄H₅IN₄
MDL Number: MFCD22570516
CAS Number: 157924-46-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR