α-Bromo-2-nitrotoluene 98%

Supplier: Ambeed
Danger

Synonyms: α-Bromo-o-nitrotoluene, 1-(Bromomethyl)-2-nitrobenzene, alpha-Bromo-2-nitrotoluene, 2-Nitrobenzyl bromide

A114145-10G A114145-25G A114145-5G
77136-162EA 9.27 USD
77136-162 77136-164 77136-166
α-Bromo-2-nitrotoluene 98%
α-Bromo-2-nitrotoluene

Precaución: for Research Use Only

Formula: C₇H₆BrNO₂
MDL Number: MFCD00007184
CAS Number: 3958-60-9
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR