(S)-Propane-1,2-diylbis(diphenylphosphine) 98%

Supplier: Ambeed
Warning

A114006-100MG A114006-1G A114006-250MG
77136-156EA 68.96 USD
77136-156 77136-158 77136-160
(S)-Propane-1,2-diylbis(diphenylphosphine) 98%
(S)-Propane-1,2-diylbis(diphenylphosphine)

Precaución: for Research Use Only

Formula: C₂₇H₂₆P₂
MW: 412.44 g/mol
MDL Number: MFCD00798604
CAS Number: 67884-33-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR