3-Chloro-4-(trifluoromethyl)phenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-Chloro-4-(trifluoromethyl)benzeneboronic acid

A113672-1G A113672-250MG A113672-5G
77136-146EA 29.71 USD
77136-146 77136-148 77136-150
3-Chloro-4-(trifluoromethyl)phenylboronic acid 97%
3-Chloro-4-(trifluoromethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₅BClF₃O₂
MW: 224.38 g/mol
MDL Number: MFCD03788417
CAS Number: 847756-88-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR