2-Chloro-4-fluoro-5-nitroanisole 97%

Supplier: Ambeed
Warning

Synonyms: 1-Chloro-5-fluoro-2-methoxy-4-nitrobenzene

A113536-100MG A113536-1G A113536-250MG A113536-25G
77136-138EA 5.15 USD
77136-138 77136-140 77136-142 77614-886
2-Chloro-4-fluoro-5-nitroanisole 97%
2-Chloro-4-fluoro-5-nitroanisole

Precaución: for Research Use Only

Formula: C₇H₅ClFNO₃
MDL Number: MFCD03425810
CAS Number: 84478-76-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR