2-Chloro-6-nitrobenzaldehyde 98%

Supplier: Ambeed
Warning

A113186-100G A113186-10G A113186-1G A113186-250MG A113186-25G A113186-5G
77136-118EA 916.71 USD
77136-118 77136-120 77136-122 77136-124 77136-126 77136-128
2-Chloro-6-nitrobenzaldehyde 98%
2-Chloro-6-nitrobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄ClNO₃
MDL Number: MFCD00007204
CAS Number: 6361-22-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR