tert-Butyl aminosulfonylcarbamate 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl sulphamoylcarbamate

A112807-100MG A112807-5G
77136-102EA 6.18 USD
77136-102 77565-976
tert-Butyl aminosulfonylcarbamate 97%
tert-Butyl aminosulfonylcarbamate

Precaución: For research use only.

Formula: C₅H₁₂N₂O₄S
MDL Number: MFCD11616268
CAS Number: 148017-28-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR