U-104 98%

Supplier: Ambeed
Warning

Synonyms: 4-(3-(4-Fluorophenyl)ureido)benzenesulphonamide

A112487-100MG A112487-10MG A112487-1MG A112487-50MG A112487-5MG
77136-092EA 113.52 USD
77136-092 77136-094 77136-096 77136-098 77136-100
U-104 98%
U-104

Precaución: for Research Use Only

Formula: C₁₃H₁₂FN₃O₃S
MW: 309.32 g/mol
MDL Number: MFCD00159265
CAS Number: 178606-66-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR