N1-(2-(Dimethylamino)ethyl)-5-methoxy-N1-methyl-N4-(4-(1-methyl-1H-indol-3-yl)pyrimidin-2-yl)benzene-1,2,4-triamine 98%

Supplier: Ambeed
Warning

Synonyms: Mutated EGFR-IN-1

A111360-10MG A111360-1MG A111360-50MG A111360-5MG A111360-1G A111360-5G A111360-25G A111360-100G
77136-076EA 8.49 USD
77136-076 77136-078 77136-080 77136-082 77549-156 77549-158 77614-862 77684-991
N1-(2-(Dimethylamino)ethyl)-5-methoxy-N1-methyl-N4-(4-(1-methyl-1H-indol-3-yl)pyrimidin-2-yl)benzene-1,2,4-triamine 98%
N1-(2-(Dimethylamino)ethyl)-5-methoxy-N1-methyl-N4-(4-(1-methyl-1H-indol-3-yl)pyrimidin-2-yl)benzene-1,2,4-triamine

Precaución: for Research Use Only

Formula: C₂₅H₃₁N₇O
MW: 445.56 g/mol
MDL Number: MFCD28167800
CAS Number: 1421372-66-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR