4-Fluoro-3-hydroxybenzonitrile 97%

Supplier: Ambeed
Warning

A111350-10G A111350-250MG A111350-25G A111350-5G A111350-1G
77136-068EA 48.41 USD
77136-068 77136-070 77136-072 77136-074 77570-636
4-Fluoro-3-hydroxybenzonitrile 97%
4-Fluoro-3-hydroxybenzonitrile

Precaución: for Research Use Only

Formula: C₇H₄FNO
MDL Number: MFCD08689519
CAS Number: 186590-04-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR