3-Formyl-6-methoxypyridine 98%

Supplier: Ambeed
Warning

Synonyms: 6-Methoxypyridine-3-carbaldehyde, 6-Methoxynicotinaldehyde, 6-Methoxypyridine-3-carboxaldehyde, 2-Methoxypyridine-5-carboxaldehyde, 6-Methoxy-3-pyridinecarboxaldehyde

A111333-100G A111333-1G A111333-250MG A111333-25G A111333-10G A111333-5G A111333-500G
77136-060EA 130.81 USD
77136-060 77136-062 77136-064 77136-066 77565-954 77592-188 77745-823
3-Formyl-6-methoxypyridine 98%
3-Formyl-6-methoxypyridine

Precaución: For research use only.

Formula: C₇H₇NO₂
MW: 137.14
MDL Number: MFCD02683446
CAS Number: 65873-72-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR