2,5-Difluoro-4-(trifluoromethyl)aniline 98%

Supplier: Ambeed
Danger

Synonyms: 4-Amino-2,5-difluorobenzotrifluoride

A111242-100MG A111242-1G A111242-250MG A111242-5G
77136-048EA 31.83 USD
77136-048 77136-050 77136-052 77136-054
2,5-Difluoro-4-(trifluoromethyl)aniline 98%
2,5-Difluoro-4-(trifluoromethyl)aniline

Precaución: for Research Use Only

Formula: C₇H₄F₅N
MW: 197.1054 g/mol
MDL Number: MFCD00190121
CAS Number: 114973-22-7
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR