2-Chloro-6-fluoro-5-methylphenylboronic acid 97%

Supplier: Ambeed
Warning

A111074-1G A111074-25G A111074-5G
77136-042EA 20.16 USD
77136-042 77136-044 77136-046
2-Chloro-6-fluoro-5-methylphenylboronic acid 97%
2-Chloro-6-fluoro-5-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₇BClFO₂
MW: 188.3917 g/mol
MDL Number: MFCD03701535
CAS Number: 352535-86-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR