Methyl 7-nitro-1H-indazole-5-carboxylate 97%

Supplier: Ambeed
Warning

A111000-100MG A111000-250MG
77136-038EA 65.78 USD
77136-038 77136-040
Methyl 7-nitro-1H-indazole-5-carboxylate 97%
Methyl 7-nitro-1H-indazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₉H₇N₃O₄
MW: 221.17 g/mol
MDL Number: MFCD22200934
CAS Number: 1823236-18-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR