(R)-(4-Benzylmorpholin-3-yl)methanol 95%

Supplier: Ambeed
Warning

A110843-1G A110843-250MG A110843-5G
77136-016EA 99.73 USD
77136-016 77136-018 77614-856
(R)-(4-Benzylmorpholin-3-yl)methanol 95%
(R)-(4-Benzylmorpholin-3-yl)methanol

Precaución: for Research Use Only

Formula: C₁₂H₁₇NO₂
MW: 207.2700 g/mol
MDL Number: MFCD06799481
CAS Number: 101376-26-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR