ABT751 98%

Supplier: Ambeed
Warning

Synonyms: N-(2-((4-Hydroxyphenyl)amino)pyridin-3-yl)-4-methoxybenzenesulphonamide, N-[2-[(4-Hydroxyphenyl)amino]-3-pyridinyl]-4-methoxybenzenesulphonamide, N-(2-((4-Hydroxyphenyl)amino)-3-pyridinyl)-4-methoxybenzenesulphonamide, ABT-751, N-[2-[(4-Hydroxyphenyl)amino]pyridin-3-yl]-4-methoxybenzenesulphonamide

A110139-10MG A110139-1MG A110139-25MG A110139-50MG A110139-5MG
77135-962EA 37.14 USD
77135-962 77135-964 77135-966 77135-968 77135-970
ABT751 98%
ABT751

Precaución: for Research Use Only

Formula: C₁₈H₁₇N₃O₄S
MW: 371.41 g/mol
MDL Number: MFCD00910291
CAS Number: 141430-65-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR