ABC294640 98%

Supplier: Ambeed
Warning

Synonyms: 1S,3R,5R,7S)-3-(4-chlorophenyl)-N-(pyridin-4-ylmethyl)adamantane-1-carboxamide

A110039-1MG A110039-5MG A110039-10MG
77135-952EA 25.47 USD
77135-952 77135-954 77684-987
ABC294640 98%
ABC294640

Precaución: for Research Use Only

Formula: C₂₃H₂₅ClN₂O
MW: 380.91 g/mol
MDL Number: MFCD22666355
CAS Number: 915385-81-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR