Benzyl (3-hydroxycyclopentyl)carbamate 98%

Supplier: Ambeed
Warning

A109831-100MG A109831-1G A109831-250MG A109831-5G
77135-942EA 24.73 USD
77135-942 77135-944 77135-946 77135-948
Benzyl (3-hydroxycyclopentyl)carbamate 98%
Benzyl (3-hydroxycyclopentyl)carbamate

Precaución: for Research Use Only

Formula: C₁₃H₁₇NO₃
MW: 235.28 g/mol
MDL Number: MFCD11848126
CAS Number: 939426-84-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR