3-Chloro-2-iodotoluene 99%

Supplier: Ambeed
Warning

Synonyms: 1-Chloro-2-iodo-3-methylbenzene

A109685-10G A109685-1G A109685-25G A109685-5G A109685-100G
77135-930EA 46.68 USD
77135-930 77135-932 77135-934 77135-936 77745-815
3-Chloro-2-iodotoluene 99%
3-Chloro-2-iodotoluene

Precaución: for Research Use Only

Formula: C₇H₆ClI
MW: 252.48 g/mol
MDL Number: MFCD00051756
CAS Number: 5100-98-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR