3-Amino-4-chlorobenzoic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-Amino-4-chlorobenzoic acid, 3-Amino-4-chlorobenzoic Acid

A109356-100G A109356-10G A109356-25G A109356-500G A109356-5G
77135-904EA 41.38 USD
77135-904 77135-906 77135-908 77135-910 77135-912
3-Amino-4-chlorobenzoic acid 97%
3-Amino-4-chlorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
MDL Number: MFCD00007671
CAS Number: 2840-28-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR