2-(9H-Carbazol-9-yl)acetic acid 95%

Supplier: Ambeed
Warning

A109091-1G A109091-5G A109091-250MG A109091-100MG
77135-870EA 176.13 USD
77135-870 77135-872 77684-982 77684-981
2-(9H-Carbazol-9-yl)acetic acid 95%
2-(9H-Carbazol-9-yl)acetic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₁NO₂
MDL Number: MFCD00439433
CAS Number: 524-80-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR