(±)-Sulfinpyrazone 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1,2-Diphenyl-4-(2-(phenylsulphinyl)ethyl)pyrazolidine-3,5-dione, sulphinpyrazone, 1,2-Diphenyl-4-[2-(phenylsulphinyl)ethyl]pyrazolidine-3,5-dione
A108887-1G
A108887-250MG
77135-858EA
6.37
USD
77135-858
77135-860
(±)-Sulfinpyrazone 98%
(±)-Sulfinpyrazone
Precaución: for Research Use Only
Formula:
C₂₃H₂₀N₂O₃S MW: 404.484 g/mol |
MDL Number:
MFCD00057279 CAS Number: 57-96-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...