(6-Bromo-1,3-benzodioxol-5-yl)methanol 95%

Supplier: Ambeed
Warning

Synonyms: (6-Bromobenzo[d][1,3]dioxol-5-yl)methanol

A108706-100MG A108706-1G A108706-250MG A108706-5G
77135-836EA 40.32 USD
77135-836 77135-838 77135-840 77745-812
(6-Bromo-1,3-benzodioxol-5-yl)methanol 95%
(6-Bromo-1,3-benzodioxol-5-yl)methanol

Precaución: for Research Use Only

Formula: C₈H₇BrO₃
MW: 231.05 g/mol
MDL Number: MFCD02082027
CAS Number: 6642-34-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR