StemRegenin 1 ≥98%

Supplier: Ambeed
Warning

Synonyms: 4-(2-((2-Benzo[b]thiphen-3-yl)-9-isopropyl-9H-purin-6-yl)amino)ethyl)phenol hydrochloride, SR1, 4-(2-((2-(Benzo[b]thiophen-3-yl)-9-isopropyl-9H-purin-6-yl)amino)ethyl)phenol

A108671-10MG A108671-1MG
77135-832EA 51.51 USD
77135-832 77135-834
StemRegenin 1 ≥98%
StemRegenin 1

Precaución: for Research Use Only

Formula: C₂₄H₂₃N₅OS
MDL Number: MFCD16987861
CAS Number: 1227633-49-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR