2-Chloro-3-nitrotoluene 98%

Supplier: Ambeed
Warning

A108405-100G A108405-10G A108405-1G A108405-25G A108405-5G
77135-818EA 191.58 USD
77135-818 77135-820 77135-822 77135-824 77135-826
2-Chloro-3-nitrotoluene 98%
2-Chloro-3-nitrotoluene

Precaución: for Research Use Only

Formula: C₇H₆ClNO₂
MDL Number: MFCD00134653
CAS Number: 3970-40-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR