(S)-Methyl 2-aminobutanoate hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A108187-100G
A108187-10G
A108187-1G
A108187-25G
A108187-500G
A108187-5G
77135-786EA
94.43
USD
77135-786
77135-788
77135-790
77135-792
77135-794
77135-796
(S)-Methyl 2-aminobutanoate hydrochloride 97%
(S)-Methyl 2-aminobutanoate hydrochloride
Precaución: for Research Use Only
Formula:
C₅H₁₂ClNO₂ MW: 153.60 g/mol |
MDL Number:
MFCD09037399 CAS Number: 56545-22-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...