4-Fluoro-3-formylphenylboronic acid 97%
Supplier: Ambeed
|
Warning
|
A107205-10G
A107205-1G
A107205-5G
77135-764EA
19.57
USD
77135-764
77135-766
77135-768
77135-770
4-Fluoro-3-formylphenylboronic acid 97%
4-Fluoro-3-formylphenylboronic acid
Precaución: for Research Use Only
Formula:
C₇H₆BFO₃ |
MDL Number:
MFCD02093074 CAS Number: 374538-01-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...