(4S)-4-[3-(Cyclopentyloxy)-4-methoxyphenyl]-pyrrolidin-2-one 98%

Supplier: Ambeed
Warning

Synonyms: (S)-(+)-Rolipram, (S)-4-(3-(Cyclopentyloxy)-4-methoxyphenyl)pyrrolidin-2-one, (S)-(+)-4-[3-(Cyclopentyloxy)-4-methoxyphenyl]pyrrolidin-2-one

A107169-100MG A107169-10MG A107169-1MG A107169-50MG A107169-5MG
77135-752EA 106.09 USD
77135-752 77135-754 77135-756 77135-758 77135-760
(4S)-4-[3-(Cyclopentyloxy)-4-methoxyphenyl]-pyrrolidin-2-one 98%
(4S)-4-[3-(Cyclopentyloxy)-4-methoxyphenyl]-pyrrolidin-2-one

Precaución: for Research Use Only

Formula: C₁₆H₂₁NO₃
MW: 275.35 g/mol
MDL Number: MFCD03093861
CAS Number: 85416-73-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR