(3-Bromobenzyl)dimethylamine 97%

Supplier: Ambeed
Warning

A107104-1G A107104-250MG A107104-25G A107104-5G
77135-742EA 126.25 USD
77135-742 77135-744 77135-746 77135-748 77135-750
(3-Bromobenzyl)dimethylamine 97%
(3-Bromobenzyl)dimethylamine

Precaución: for Research Use Only

Formula: C₉H₁₂BrN
MDL Number: MFCD04117937
CAS Number: 4885-18-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR