Amlodipine maleate 98%

Supplier: Ambeed
Warning

Synonyms: 3-Ethyl 5-methyl 2-((2-aminoethoxy)methyl)-4-(2-chlorophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate maleate

A107087-100MG A107087-10G A107087-1G A107087-1MG A107087-250MG A107087-25G A107087-5G
77135-728EA 7.21 USD
77135-728 77135-730 77135-732 77135-734 77135-736 77135-738 77135-740
Amlodipine maleate 98%
Amlodipine maleate

Precaución: for Research Use Only

Formula: C₂₄H₂₉ClN₂O₉
MW: 524.94 g/mol
MDL Number: MFCD02259457
CAS Number: 88150-47-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR