tert-Butyl 3-(phenylamino)azetidine-1-carboxylate 97%

Supplier: Ambeed
Warning

A106849-100MG A106849-1G A106849-250MG
77135-712EA 78.51 USD
77135-712 77135-714 77135-716
tert-Butyl 3-(phenylamino)azetidine-1-carboxylate 97%
tert-Butyl 3-(phenylamino)azetidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₂₀N₂O₂
MW: 248.32 g/mol
MDL Number: MFCD19471441
CAS Number: 1285278-84-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR