Boc-(R)-3-amino-4-(4-bromophenyl)butyric acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(R)-4-(4-Bromophenyl)-3-((tert-butoxycarbonyl)amino)butanoic acid
A104883-100MG
A104883-1G
A104883-250MG
77135-660EA
29.71
USD
77135-660
77135-662
77135-664
Boc-(R)-3-amino-4-(4-bromophenyl)butyric acid 98%
Boc-(R)-3-amino-4-(4-bromophenyl)butyric acid
Precaución: for Research Use Only
Formula:
C₁₅H₂₀BrNO₄ MW: 358.24 g/mol |
MDL Number:
MFCD01860917 CAS Number: 331763-75-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...