R406 ≥98%

Supplier: Ambeed
Warning

Synonyms: 6-((5-Fluoro-2-((3,4,5-trimethoxyphenyl)amino)pyrimidin-4-yl)amino)-2,2-dimethyl-2H-pyrido[3,2-b][1,4]oxazin-3(4H)-one benzenesulphonate

A104426-10MG A104426-1MG A104426-5MG
77135-638EA 24.41 USD
77135-638 77135-640 77135-642
R406 ≥98%
R406

Precaución: for Research Use Only

Formula: C₂₈H₂₉FN₆O₈S
MW: 628.63 g/mol
MDL Number: MFCD18385012
CAS Number: 841290-81-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR