4-Bromo-2-chloro-5-(difluoromethyl)pyridine 95%

Supplier: Ambeed
Danger

A104180-1G A104180-100MG
77135-636EA 750.06 USD
77135-636 77684-947
4-Bromo-2-chloro-5-(difluoromethyl)pyridine 95%
4-Bromo-2-chloro-5-(difluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₃BrClF₂N
MW: 242.45 g/mol
MDL Number: MFCD25477654
CAS Number: 1471259-57-0
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR