Methotrexate 99% (contain ~10% water)
Supplier: Ambeed
|
Warning
|
Synonyms:
(S)-2-(4-(((2,4-Diaminopteridin-6-yl)methyl)(methyl)amino)benzamido)pentanedioic acid
A103612-10G
A103612-1G
A103612-1MG
A103612-25G
A103612-5G
A103612-100G
77135-614EA
68.96
USD
77135-614
77135-616
77135-618
77135-620
77135-622
77684-942
Methotrexate 99% (contain ~10% water)
Methotrexate
Precaución: for Research Use Only
Formula:
C₂₀H₂₂N₈O₅ MW: 454.44 g/mol |
MDL Number:
MFCD00150847 CAS Number: 59-05-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...