(2-(((1-Chloroethoxy)carbonyl)(methyl)amino)pyridin-3-yl)methyl 2-((tert-butoxycarbonyl)(methyl)amino)acetate 95%
Supplier: Ambeed
|
Warning
|
A103562-1G
A103562-250MG
A103562-5G
77135-610EA
16.48
USD
77135-610
77135-612
77684-941
(2-(((1-Chloroethoxy)carbonyl)(methyl)amino)pyridin-3-yl)methyl 2-((tert-butoxycarbonyl)(methyl)amino)acetate 95%
(2-(((1-Chloroethoxy)carbonyl)(methyl)amino)pyridin-3-yl)methyl 2-((tert-butoxycarbonyl)(methyl)amino)acetate
Precaución: for Research Use Only
Formula:
C₁₈H₂₆ClN₃O₆ MW: 415.86 g/mol |
MDL Number:
MFCD30184953 CAS Number: 338990-31-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...