Dimethyl cyclopentane-1,3-dicarboxylate 95%

Supplier: Ambeed
Warning

A103244-100G A103244-100MG A103244-10G A103244-1G A103244-250MG A103244-25G A103244-5G
77135-588EA 521.97 USD
77135-588 77135-590 77135-592 77135-594 77135-596 77135-598 77135-600
Dimethyl cyclopentane-1,3-dicarboxylate 95%
Dimethyl cyclopentane-1,3-dicarboxylate

Precaución: for Research Use Only

Formula: C₉H₁₄O₄
MW: 186.20 g/mol
MDL Number: MFCD01735429
CAS Number: 2435-36-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR