1-Chloro-3-methyl-5-nitrobenzene 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Chloro-5-nitrotoluene
A103209-100G
A103209-10G
A103209-1G
A103209-25G
A103209-5G
77135-578EA
97.85
USD
77135-578
77135-580
77135-582
77135-584
77135-586
1-Chloro-3-methyl-5-nitrobenzene 98%
1-Chloro-3-methyl-5-nitrobenzene
Precaución: for Research Use Only
Formula:
C₇H₆ClNO₂ MW: 171.58 g/mol |
MDL Number:
MFCD09955417 CAS Number: 16582-38-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...