(4-Chloro-7H-pyrrolo[2,3-d]pyrimidin-7-yl)methyl pivalate 97%

Supplier: Ambeed
Warning

A102895-1G A102895-5G
77135-574EA 12.74 USD
77135-574 77135-576
(4-Chloro-7H-pyrrolo[2,3-d]pyrimidin-7-yl)methyl pivalate 97%
(4-Chloro-7H-pyrrolo[2,3-d]pyrimidin-7-yl)methyl pivalate

Precaución: for Research Use Only

Formula: C₁₂H₁₄ClN₃O₂
MW: 267.71 g/mol
MDL Number: MFCD29060065
CAS Number: 1146629-75-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR