6-Bromo-3-fluoropyridin-2-amine 98%

Supplier: Ambeed
Warning

A102500-100MG A102500-1G A102500-250MG A102500-5G
77135-550EA 40.17 USD
77135-550 77135-552 77135-554 77135-556
6-Bromo-3-fluoropyridin-2-amine 98%
6-Bromo-3-fluoropyridin-2-amine

Precaución: for Research Use Only

Formula: C₅H₄BrFN₂
MDL Number: MFCD28368857
CAS Number: 1379457-78-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR