Ethyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[b]thiophene-2-carboxylate 97%

Supplier: Ambeed
Warning

A102391-100MG A102391-1G A102391-250MG
77135-544EA 36.08 USD
77135-544 77135-546 77135-548
Ethyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[b]thiophene-2-carboxylate 97%
Ethyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[b]thiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₇H₂₁BO₄S
MW: 332.22 g/mol
MDL Number: MFCD24448814
CAS Number: 2377609-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR