5-Bromo-2-hydroxyisonicotinaldehyde 97%

Supplier: Ambeed
Warning

A101919-100MG A101919-250MG A101919-1G
77135-518EA 22.66 USD
77135-518 77135-520 77592-074
5-Bromo-2-hydroxyisonicotinaldehyde 97%
5-Bromo-2-hydroxyisonicotinaldehyde

Precaución: for Research Use Only

Formula: C₆H₄BrNO₂
MDL Number: MFCD16606520
CAS Number: 1227562-37-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR