N-(2-(Dimethylamino)ethyl)-12-oxo-12H-benzo[g]pyrido[2,1-b]quinazoline-4-carboxamide 98%

Supplier: Ambeed
Warning

Synonyms: BMH-21

A101477-10MG A101477-1MG A101477-25MG A101477-5MG
77135-494EA 40.32 USD
77135-494 77135-496 77135-498 77135-500
N-(2-(Dimethylamino)ethyl)-12-oxo-12H-benzo[g]pyrido[2,1-b]quinazoline-4-carboxamide 98%
N-(2-(Dimethylamino)ethyl)-12-oxo-12H-benzo[g]pyrido[2,1-b]quinazoline-4-carboxamide

Precaución: for Research Use Only

Formula: C₂₁H₂₀N₄O₂
MW: 360.41 g/mol
MDL Number: MFCD27225377
CAS Number: 896705-16-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR