Benzyl nicotinate 97%

Supplier: Ambeed
Warning

A100203-100G A100203-10G A100203-5G A100203-500G A100203-25G
77135-414EA 48.41 USD
77135-414 77135-416 77135-418 77570-004 77614-724
Benzyl nicotinate 97%
Benzyl nicotinate

Precaución: for Research Use Only

Formula: C₁₃H₁₁NO₂
MDL Number: MFCD00023584
CAS Number: 94-44-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR