(S)-(-)-XylBINAP 98%

Supplier: Ambeed
Warning

Synonyms: (S)-2,2'-Bis(bis(3,5-dimethylphenyl)phosphino)-1,1'-binaphthalene, (S)-(-)-2,2'-Bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl

A999826-100MG A999826-1G A999826-250MG A999826-5G
77119-136EA 11.33 USD
77119-136 77119-138 77119-140 77119-142
(S)-(-)-XylBINAP 98%
(S)-(-)-XylBINAP

Precaución: for Research Use Only

Formula: C₅₂H₄₈P₂
MDL Number: MFCD01630821
CAS Number: 135139-00-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR