4-Chloro-5-iodo-2-(trifluoromethyl)pyridine 95%

Supplier: Ambeed
Warning

A999119-100MG A999119-1G A999119-5G
77119-130EA 38.11 USD
77119-130 77119-132 77119-134
4-Chloro-5-iodo-2-(trifluoromethyl)pyridine 95%
4-Chloro-5-iodo-2-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₂ClF₃IN
MW: 307.43 g/mol
MDL Number: MFCD24369746
CAS Number: 1027818-88-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR